Assay Details
| Gene Symbol: ARF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001662.3 | NP_001653.1 | 1 - 2 | 213 | 92 | |
| GenBank mRNA | AF493884.1 | - | 1 - 2 | 66 | 92 | |
| AK297026.1 | - | 1 - 2 | 118 | 92 | ||
| AK307297.1 | - | 1 - 2 | 187 | 92 | ||
| AK312107.1 | - | 1 - 2 | 118 | 92 | ||
| BC003043.1 | - | 1 - 2 | 103 | 2822973 | 92 | |
| BC033104.1 | - | 1 - 2 | 160 | 4862014 | 92 | |
| BT007087.1 | - | 1 - 2 | 66 | 92 | ||
| CN259625.1 | - | 1 - 2 | 230 | 92 | ||
| M57567.1 | - | 1 - 2 | 120 | 92 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003341.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07012040.hg.1 | ENST00000000233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012040.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012040.hg.1 | TC07003341.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012048.hg.1 | ENST00000000233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012048.hg.1 | ENST00000467281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012048.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012048.hg.1 | TC07003341.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012044.hg.1 | ENST00000000233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012044.hg.1 | ENST00000467281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012044.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012044.hg.1 | TC07003341.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012045.hg.1 | ENST00000000233.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012045.hg.1 | ENST00000467281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012045.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012045.hg.1 | TC07003341.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000459680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000459680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000459680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000459680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000459680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000459680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000459680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000459680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013456.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700198795.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198795.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198795.hg.1 | ENST00000459680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198795.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198795.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198795.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198795.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700198795.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198795.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198795.hg.1 | ENST00000459680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198795.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198795.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198795.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198795.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700198795.hg.1 | TC0700013456.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700198790.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198790.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198790.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198790.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198790.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198790.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700198790.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198790.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198790.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198790.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198790.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198790.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700198790.hg.1 | TC0700013456.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700198791.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198791.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198791.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198791.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198791.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198791.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700198791.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198791.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198791.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198791.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198791.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198791.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700198791.hg.1 | TC0700013456.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700198792.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198792.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198792.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198792.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198792.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198792.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700198792.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198792.hg.1 | ENST00000415666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198792.hg.1 | ENST00000463733.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198792.hg.1 | ENST00000467281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198792.hg.1 | ENST00000489673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198792.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700198792.hg.1 | TC0700013456.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700198786.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198786.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700198786.hg.1 | ENST00000000233.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700198786.hg.1 | NM_001662.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700198786.hg.1 | TC0700013456.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 177382 | - | - |

ARF5
Genomic Map