Assay Details
Target Gene Details
Entrez Gene ID: | 7941 |
Gene Name: | phospholipase A2 group VII |
Gene Aliases: |
LDL-PLA2, LP-PLA2, PAFAD, PAFAH |
Location: |
Chr.6:46700954-46735836 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G7 | NM_001168357.1 | 12 | 1435 | NP_001161829.1 |
| NM_005084.3 | 12 | 1526 | NP_005075.3 | |
| XM_005249408.4 | 12 | 1510 | XP_005249465.1 | |
| AK290381.1 | 12 | 1485 | ||
| BC038452.1 | 12 | 1435 | AAH38452.1 | |
| U20157.1 | 12 | 1418 | AAC50126.1 | |
| U24577.1 | 12 | 1473 | AAB04170.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523148 | Chr.6:46702199 - 46705206 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map