Assay Details
Target Gene Details
Entrez Gene ID: | 1756 |
Gene Name: | dystrophin |
Gene Aliases: |
BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, MRX85 |
Location: |
Chr.X:31119219-33339609 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 86 - Exon 86 |
More Information
Set Membership: |
|

Genomic Map