Assay Details
Target Gene Details
Entrez Gene ID: | 7273 |
Gene Name: | titin |
Gene Aliases: |
CMD1G, CMH9, CMPD4, EOMFC, HMERF, LGMD2J, MYLK5, TMD |
Location: |
Chr.2:178525989-178807423 on Build GRCh38 |
Assay Gene Location: | Within Exon 359 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTN | NM_001256850.1 | 308 | 98747 | NP_001243779.1 |
| NM_001267550.2 | 358 | 103670 | NP_001254479.2 | |
| NM_003319.4 | 186 | 76475 | NP_003310.4 | |
| NM_133378.4 | 307 | 95966 | NP_596869.4 | |
| NM_133432.3 | 187 | 76850 | NP_597676.3 | |
| NM_133437.4 | 187 | 77051 | NP_597681.4 | |
| XM_017004819.1 | 354 | 102563 | XP_016860308.1 | |
| XM_017004820.1 | 331 | 97961 | XP_016860309.1 | |
| XM_017004821.1 | 331 | 97958 | XP_016860310.1 | |
| XM_017004822.1 | 269 | 95000 | XP_016860311.1 | |
| XM_017004823.1 | 187 | 76616 | XP_016860312.1 | |
| X69490.1 | 20 | 9422 | CAA49245.1 | |
| X90568.1 | 186 | 76382 | CAA62188.1 |
Target Gene Details
Entrez Gene ID: | 100506866 |
Gene Name: | TTN antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:178522827-178620217 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTN-AS1 | NR_038271.1 | |||
| NR_038272.1 | ||||
| AK093733.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834468 | Chr.2:178486389 - 178668898 on Build GRCh38 | Gain |
|
| nsv834467 | Chr.2:178393413 - 178566238 on Build GRCh38 | Loss |
|
| nsv522036 | Chr.2:178531033 - 178533782 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map