Assay Details
Target Gene Details
Entrez Gene ID: | 5213 |
Gene Name: | phosphofructokinase, muscle |
Gene Aliases: |
ATP-PFK, GSD7, PFK-1, PFK1, PFKA, PFKX, PPP1R122 |
Location: |
Chr.12:48105278-48146404 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759898 | Chr.12:48035919 - 48431716 on Build GRCh38 | Gain+Loss |
|
| nsv508673 | Chr.12:48096003 - 48173355 on Build GRCh38 | Deletion |
|
| nsv558790 | Chr.12:48135385 - 48146105 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map