Assay Details
Target Gene Details
Entrez Gene ID: | 2259 |
Gene Name: | fibroblast growth factor 14 |
Gene Aliases: |
FGF-14, FHF-4, FHF4, SCA27 |
Location: |
Chr.13:101720855-102402428 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832702 | Chr.13:101621711 - 101795212 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map