Assay Details
Target Gene Details
Entrez Gene ID: | 23512 |
Gene Name: | SUZ12 polycomb repressive complex 2 subunit |
Gene Aliases: |
CHET9, JJAZ1 |
Location: |
Chr.17:31937010-32001045 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUZ12 | NM_001321207.1 | 12 | 1688 | NP_001308136.1 |
| NM_015355.3 | 13 | 1757 | NP_056170.2 | |
| XM_006721794.3 | 11 | 1199 | XP_006721857.1 | |
| XM_017024409.1 | 12 | 1413 | XP_016879898.1 | |
| AK074333.1 | 8 | 994 | ||
| AK290014.1 | 13 | 1728 | ||
| BC015704.1 | 13 | 1599 | AAH15704.1 | |
| D63881.2 | 13 | 1714 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062194 | Chr.17:31755256 - 32041289 on Build GRCh38 | Gain |
|
| esv2758684 | Chr.17:31894702 - 32210963 on Build GRCh38 | Gain |
|
| nsv1067240 | Chr.17:31892481 - 32005581 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map