Assay Details
Target Gene Details
Entrez Gene ID: | 1649 |
Gene Name: | DNA damage inducible transcript 3 |
Gene Aliases: |
CEBPZ, CHOP, CHOP-10, CHOP10, GADD153 |
Location: |
Chr.12:57516588-57520517 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDIT3 | NM_001195053.1 | 4 | 528 | NP_001181982.1 |
| NM_001195054.1 | 4 | 475 | NP_001181983.1 | |
| NM_001195055.1 | 4 | 452 | NP_001181984.1 | |
| NM_001195056.1 | 3 | 638 | NP_001181985.1 | |
| NM_001195057.1 | 4 | 357 | NP_001181986.1 | |
| NM_004083.5 | 4 | 371 | NP_004074.2 | |
| AA054097.1 | ||||
| AA476561.1 | 4 | 378 | ||
| AK316581.1 | 3 | 632 | ||
| AV729744.1 | 4 | 357 | ||
| AY461580.1 | 4 | 530 | ||
| BC003637.2 | 4 | 372 | AAH03637.1 | |
| BM464290.1 | 4 | 361 | ||
| BT006691.1 | 2 | 191 | AAP35337.1 | |
| BU664077.1 | 4 | 469 | ||
| CN482428.1 | 4 | 335 | ||
| S40706.1 | 4 | 362 | AAB22646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759901 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain |
|
| nsv559034 | Chr.12:57466149 - 57618450 on Build GRCh38 | Loss |
|
| dgv2651n54 | Chr.12:57450266 - 57552694 on Build GRCh38 | Loss |
|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map