Assay Details
Target Gene Details
Entrez Gene ID: | 51592 |
Gene Name: | tripartite motif containing 33 |
Gene Aliases: |
ECTO, PTC7, RFG7, TF1G, TIF1G, TIF1GAMMA, TIFGAMMA |
Location: |
Chr.1:114392777-114511160 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM33 | NM_015906.3 | 20 | 6291 | NP_056990.3 |
| NM_033020.2 | 19 | 6240 | NP_148980.2 | |
| XM_005270936.3 | 21 | 6284 | XP_005270993.1 | |
| XM_005270937.3 | 20 | 6233 | XP_005270994.1 | |
| XM_011541568.2 | 21 | 6281 | XP_011539870.1 | |
| XM_017001452.1 | 18 | 5566 | XP_016856941.1 | |
| XM_017001453.1 | 17 | 5515 | XP_016856942.1 | |
| AL080216.1 | 1 | 166 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28734 | Chr.1:114376708 - 115005298 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map