Assay Details
Target Gene Details
Entrez Gene ID: | 4803 |
Gene Name: | nerve growth factor |
Gene Aliases: |
Beta-NGF, HSAN5, NGFB |
Location: |
Chr.1:115285915-115338253 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NGF | NM_002506.2 | 3 | 645 | NP_002497.2 |
| XM_006710663.3 | 2 | 538 | XP_006710726.1 | |
| XM_011541518.2 | 3 | 3683 | XP_011539820.1 | |
| AF411526.1 | 2 | 549 | AAL05874.1 | |
| BC032517.2 | 3 | 650 | AAH32517.2 | |
| BC126148.1 | 2 | 572 | ||
| BC126150.1 | 2 | 572 | ||
| BE045524.1 | ||||
| BT019733.1 | 1 | 476 | AAV38538.1 | |
| CR541855.1 | 1 | 476 | CAG46653.1 | |
| X52599.1 | 3 | 645 | CAA36832.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34370 | Chr.1:115181074 - 115625337 on Build GRCh38 | Gain |
|
| esv26121 | Chr.1:115020681 - 115560070 on Build GRCh38 | Loss |
|
| nsv818389 | Chr.1:115185327 - 115526418 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map