Assay Details
Target Gene Details
Entrez Gene ID: | 101927055 |
Gene Name: | uncharacterized LOC101927055 |
Gene Aliases: |
- |
Location: |
Chr.2:178776926-178779963 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927055 | NR_120594.1 | 1 | 1025 | |
| AK123298.1 | 1 | 1025 |
Target Gene Details
Entrez Gene ID: | 7273 |
Gene Name: | titin |
Gene Aliases: |
CMD1G, CMH9, CMPD4, EOMFC, HMERF, LGMD2J, MYLK5, TMD |
Location: |
Chr.2:178525989-178807423 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 24 - Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTN | NM_001256850.1 | NP_001243779.1 | ||
| NM_001267550.2 | NP_001254479.2 | |||
| NM_003319.4 | NP_003310.4 | |||
| NM_133378.4 | NP_596869.4 | |||
| NM_133379.4 | NP_596870.2 | |||
| NM_133432.3 | NP_597676.3 | |||
| NM_133437.4 | NP_597681.4 | |||
| XM_017004819.1 | XP_016860308.1 | |||
| XM_017004820.1 | XP_016860309.1 | |||
| XM_017004821.1 | XP_016860310.1 | |||
| XM_017004822.1 | XP_016860311.1 | |||
| XM_017004823.1 | XP_016860312.1 | |||
| X90568.1 | CAA62188.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834470 | Chr.2:178690425 - 178863999 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map