Assay Details
Target Gene Details
Entrez Gene ID: | 7273 |
Gene Name: | titin |
Gene Aliases: |
CMD1G, CMH9, CMPD4, EOMFC, HMERF, LGMD2J, MYLK5, TMD |
Location: |
Chr.2:178525989-178807423 on Build GRCh38 |
Assay Gene Location: | Within Exon 364 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTN | NM_001256850.1 | 313 | 103934 | NP_001243779.1 |
| NM_001267550.2 | 363 | 108857 | NP_001254479.2 | |
| NM_003319.4 | 191 | 81662 | NP_003310.4 | |
| NM_133378.4 | 312 | 101153 | NP_596869.4 | |
| NM_133432.3 | 192 | 82037 | NP_597676.3 | |
| NM_133437.4 | 192 | 82238 | NP_597681.4 | |
| XM_017004819.1 | 359 | 107750 | XP_016860308.1 | |
| XM_017004820.1 | 336 | 103148 | XP_016860309.1 | |
| XM_017004821.1 | 336 | 103145 | XP_016860310.1 | |
| XM_017004822.1 | 274 | 100187 | XP_016860311.1 | |
| XM_017004823.1 | 192 | 81803 | XP_016860312.1 | |
| AL713647.1 | 6 | 4355 | CAD28458.1 | |
| BC107797.1 | 3 | 1196 | AAI07798.1 | |
| X69490.1 | 25 | 14609 | CAA49245.1 | |
| X90568.1 | 191 | 81569 | CAA62188.1 |
Target Gene Details
Entrez Gene ID: | 100506866 |
Gene Name: | TTN antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:178522827-178620217 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTN-AS1 | NR_038271.1 | |||
| NR_038272.1 | ||||
| AK093733.1 | ||||
| DB229458.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834468 | Chr.2:178486389 - 178668898 on Build GRCh38 | Gain |
|
| nsv834467 | Chr.2:178393413 - 178566238 on Build GRCh38 | Loss |
|
| nsv954437 | Chr.2:178523774 - 178531073 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72629796] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map