Assay Details
Target Gene Details
Entrez Gene ID: | 9231 |
Gene Name: | discs large MAGUK scaffold protein 5 |
Gene Aliases: |
LP-DLG, P-DLG5, PDLG |
Location: |
Chr.10:77790787-77926590 on Build GRCh38 |
Assay Gene Location: | Within Exon 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLG5 | NM_004747.3 | 32 | 6517 | NP_004738.3 |
| XM_005270276.4 | 32 | 6471 | XP_005270333.1 | |
| XM_006718056.3 | 31 | 5460 | XP_006718119.1 | |
| XM_011540341.2 | 32 | 6641 | XP_011538643.1 | |
| XM_011540342.1 | 32 | 6219 | XP_011538644.1 | |
| XM_011540344.2 | 31 | 6126 | XP_011538646.1 | |
| XM_011540345.1 | 31 | 6251 | XP_011538647.1 | |
| XM_011540347.2 | 28 | 5697 | XP_011538649.1 | |
| XM_017016913.1 | 32 | 6180 | XP_016872402.1 | |
| XM_017016914.1 | 32 | 6510 | XP_016872403.1 | |
| XM_017016915.1 | 28 | 6381 | XP_016872404.1 | |
| AB011155.2 | 32 | 6547 | ||
| AK055265.1 | 1 | 1527 | ||
| BC073996.1 | 18 | 3227 | AAH73996.1 | |
| BC146794.1 | 32 | 6547 | ||
| BX649131.1 | 27 | 4625 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv929n100 | Chr.10:77596297 - 77855405 on Build GRCh38 | Gain |
|
| esv34251 | Chr.10:77609516 - 77833713 on Build GRCh38 | Gain |
|
| esv3623882 | Chr.10:77784321 - 77835624 on Build GRCh38 | Gain |
|
| esv3623881 | Chr.10:77609729 - 77835435 on Build GRCh38 | Gain |
|
| esv2759769 | Chr.10:77351633 - 77833713 on Build GRCh38 | Gain+Loss |
|
| esv34062 | Chr.10:77772218 - 77807341 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map