Assay Details
Target Gene Details
Entrez Gene ID: | 51592 |
Gene Name: | tripartite motif containing 33 |
Gene Aliases: |
ECTO, PTC7, RFG7, TF1G, TIF1G, TIF1GAMMA, TIFGAMMA |
Location: |
Chr.1:114392777-114511160 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM33 | NM_015906.3 | 20 | 6932 | NP_056990.3 |
| NM_033020.2 | 19 | 6881 | NP_148980.2 | |
| XM_005270936.3 | 21 | 6925 | XP_005270993.1 | |
| XM_005270937.3 | 20 | 6874 | XP_005270994.1 | |
| XM_011541568.2 | 21 | 6922 | XP_011539870.1 | |
| XM_017001452.1 | 18 | 6207 | XP_016856941.1 | |
| XM_017001453.1 | 17 | 6156 | XP_016856942.1 | |
| AL080216.1 | 1 | 807 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28734 | Chr.1:114376708 - 115005298 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs148085403] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map