Assay Details
Target Gene Details
Entrez Gene ID: | 5311 |
Gene Name: | polycystin 2, transient receptor potential cation channel |
Gene Aliases: |
APKD2, PC2, PKD4, Pc-2, TRPP2 |
Location: |
Chr.4:88007647-88077779 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKD2 | NM_000297.3 | 2 | 722 | NP_000288.1 |
| XM_011532028.2 | 2 | 715 | XP_011530330.1 | |
| BC112261.1 | 2 | 722 | AAI12262.1 | |
| BC112263.1 | 2 | 722 | AAI12264.1 | |
| U50928.1 | 2 | 701 | AAC50520.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2665331 | Chr.4:87937174 - 88153583 on Build GRCh38 | Deletion |
|
| esv2759266 | Chr.4:87997399 - 88291967 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map