Assay Details
Target Gene Details
Entrez Gene ID: | 51592 |
Gene Name: | tripartite motif containing 33 |
Gene Aliases: |
ECTO, PTC7, RFG7, TF1G, TIF1G, TIF1GAMMA, TIFGAMMA |
Location: |
Chr.1:114392777-114511160 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM33 | NM_015906.3 | NP_056990.3 | ||
| NM_033020.2 | NP_148980.2 | |||
| XM_005270936.3 | XP_005270993.1 | |||
| XM_005270937.3 | XP_005270994.1 | |||
| XM_011541568.2 | XP_011539870.1 | |||
| XM_017001452.1 | XP_016856941.1 | |||
| XM_017001453.1 | XP_016856942.1 | |||
| AB029036.1 | ||||
| AF119043.1 | AAD17259.1 | |||
| AF220136.1 | AAG53509.1 | |||
| AF220137.1 | AAG53510.1 | |||
| AJ132948.1 | CAB55313.1 | |||
| AK023434.1 | ||||
| AK057487.1 | ||||
| AK301997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28734 | Chr.1:114376708 - 115005298 on Build GRCh38 | Loss |
|
| esv3587262 | Chr.1:114396576 - 114425946 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map