Assay Details
Target Gene Details
Entrez Gene ID: | 7941 |
Gene Name: | phospholipase A2 group VII |
Gene Aliases: |
LDL-PLA2, LP-PLA2, PAFAD, PAFAH |
Location: |
Chr.6:46700954-46735836 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G7 | NM_001168357.1 | NP_001161829.1 | ||
| NM_005084.3 | NP_005075.3 | |||
| XM_005249408.4 | XP_005249465.1 | |||
| AK290381.1 | ||||
| BC038452.1 | AAH38452.1 | |||
| U20157.1 | AAC50126.1 | |||
| U24577.1 | AAB04170.1 |
More Information
Set Membership: |
|

Genomic Map