Assay Details
Target Gene Details
Entrez Gene ID: | 7200 |
Gene Name: | thyrotropin releasing hormone |
Gene Aliases: |
Pro-TRH, TRF |
Location: |
Chr.3:129974393-129977938 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRH | NM_007117.4 | NP_009048.1 | ||
| AK313473.1 | ||||
| BC069375.1 | AAH69375.1 | |||
| BC074888.2 | AAH74888.1 | |||
| BC074889.2 | AAH74889.1 | |||
| BC110515.2 | AAI10516.1 | |||
| BC110516.2 | AAI10517.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4860n100 | Chr.3:129963200 - 130089956 on Build GRCh38 | Gain |
|
| esv2759177 | Chr.3:129940624 - 130221139 on Build GRCh38 | Gain+Loss |
|
| nsv460853 | Chr.3:129961523 - 129990509 on Build GRCh38 | Loss |
|
| nsv1012843 | Chr.3:129957739 - 130166820 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map