Assay Details
Target Gene Details
Entrez Gene ID: | 657 |
Gene Name: | bone morphogenetic protein receptor type 1A |
Gene Aliases: |
10q23del, ACVRLK3, ALK3, CD292, SKR5 |
Location: |
Chr.10:86755786-86927969 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BMPR1A | NM_004329.2 | NP_004320.2 | ||
| XM_011540103.2 | XP_011538405.1 | |||
| XM_011540104.2 | XP_011538406.1 | |||
| AK291764.1 | ||||
| BC028383.1 | AAH28383.1 | |||
| BM819272.1 | ||||
| Z22535.1 | CAA80257.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759774 | Chr.10:86755301 - 87550005 on Build GRCh38 | Gain+Loss |
|
| nsv7481 | Chr.10:86827171 - 86849043 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map