Assay Details
Target Gene Details
Entrez Gene ID: | 1642 |
Gene Name: | damage specific DNA binding protein 1 |
Gene Aliases: |
DDBA, UV-DDB1, XAP1, XPCE, XPE, XPE-BF |
Location: |
Chr.11:61299447-61333212 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDB1 | NM_001923.4 | 27 | 4129 | NP_001914.3 |
| AL831958.1 | 16 | 3798 | ||
| BC011686.2 | 27 | 3958 | AAH11686.1 | |
| BC050530.1 | 27 | 4022 | AAH50530.1 | |
| BC051764.2 | 27 | 4012 | AAH51764.1 | |
| CD242268.1 | 1 | 261 | ||
| L40326.1 | 27 | 3923 | AAA62838.1 | |
| U18299.1 | 27 | 3985 | AAC50349.1 | |
| U32986.1 | 27 | 3994 | AAA88883.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759831 | Chr.11:61147888 - 61313185 on Build GRCh38 | Gain+Loss |
|
| nsv428259 | Chr.11:61049513 - 61356164 on Build GRCh38 | Gain+Loss |
|
| nsv509415 | Chr.11:61262472 - 61315210 on Build GRCh38 | Insertion |
|
| nsv832177 | Chr.11:61185118 - 61356164 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map