Assay Details
Target Gene Details
Entrez Gene ID: | 2581 |
Gene Name: | galactosylceramidase |
Gene Aliases: |
- |
Location: |
Chr.14:87933014-87993665 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALC | NM_000153.3 | NP_000144.2 | ||
| NM_001201401.1 | NP_001188330.1 | |||
| NM_001201402.1 | NP_001188331.1 | |||
| XM_011536618.2 | XP_011534920.1 | |||
| AK294040.1 | ||||
| AK296530.1 | ||||
| AK302519.1 | ||||
| AK302683.1 | ||||
| AK302956.1 | ||||
| BC036518.2 | AAH36518.1 | |||
| D25283.1 | BAA04971.1 | |||
| H80485.1 | ||||
| L23116.1 | AAA16645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2665905 | Chr.14:87925147 - 87956869 on Build GRCh38 | Deletion |
|
| nsv514759 | Chr.14:87933687 - 87948247 on Build GRCh38 | Gain+Loss |
|
| nsv9165 | Chr.14:87934624 - 87945563 on Build GRCh38 | Gain |
|
| esv2760008 | Chr.14:87924772 - 87963950 on Build GRCh38 | Loss |
|
| nsv565506 | Chr.14:87928587 - 87948259 on Build GRCh38 | Loss |
|
| nsv1047065 | Chr.14:87923196 - 87956225 on Build GRCh38 | Loss |
|
| nsv442353 | Chr.14:87932027 - 87956221 on Build GRCh38 | Loss |
|
| dgv1953n100 | Chr.14:87932015 - 87949156 on Build GRCh38 | Loss |
|
| dgv672e212 | Chr.14:87932014 - 87960402 on Build GRCh38 | Loss |
|
| esv3635267 | Chr.14:87833153 - 87954098 on Build GRCh38 | Gain |
|
| dgv3840n54 | Chr.14:87923791 - 87961245 on Build GRCh38 | Loss |
|
| esv3635268 | Chr.14:87925163 - 87956836 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map