Assay Details
Target Gene Details
Entrez Gene ID: | 2796 |
Gene Name: | gonadotropin releasing hormone 1 |
Gene Aliases: |
GNRH, GRH, LHRH, LNRH |
Location: |
Chr.8:25419258-25425040 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNRH1 | NM_000825.3 | NP_000816.4 | ||
| NM_001083111.1 | NP_001076580.1 | |||
| CB243067.1 | ||||
| M12578.1 | AAA35916.1 | |||
| X01059.1 | CAA25526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759606 | Chr.8:25411509 - 25564618 on Build GRCh38 | Loss |
|
| dgv7148n100 | Chr.8:25410448 - 25473361 on Build GRCh38 | Gain |
|
| nsv6125 | Chr.8:25410366 - 25444678 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map