Assay Details
Target Gene Details
Entrez Gene ID: | 5071 |
Gene Name: | parkin RBR E3 ubiquitin protein ligase |
Gene Aliases: |
AR-JP, LPRS2, PDJ, PRKN |
Location: |
Chr.6:161347417-162727802 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARK2 | NM_004562.2 | 12 | 2920 | NP_004553.2 |
| NM_013987.2 | 11 | 2836 | NP_054642.2 | |
| NM_013988.2 | 9 | 2473 | NP_054643.2 | |
| XM_011535863.1 | 12 | 2917 | XP_011534165.1 | |
| XM_017010908.1 | 12 | 3834 | XP_016866397.1 | |
| XM_017010909.1 | 11 | 2714 | XP_016866398.1 | |
| AB009973.1 | 12 | 2887 | BAA25751.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830854 | Chr.6:161318894 - 161506483 on Build GRCh38 | Gain |
|
| nsv1029727 | Chr.6:161186648 - 161393263 on Build GRCh38 | Gain |
|
| nsv1031234 | Chr.6:161255736 - 161829694 on Build GRCh38 | Gain |
|
| nsv1026543 | Chr.6:160306836 - 161585804 on Build GRCh38 | Gain |
|
| esv3576244 | Chr.6:161272053 - 161395540 on Build GRCh38 | Gain |
|
| esv3576243 | Chr.6:161178374 - 161396606 on Build GRCh38 | Gain |
|
| esv2752091 | Chr.6:161171068 - 161434753 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map