Assay Details
Target Gene Details
Entrez Gene ID: | 114299 |
Gene Name: | paralemmin 2 |
Gene Aliases: |
AKAP2 |
Location: |
Chr.9:109640788-109951476 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PALM2 | NM_001037293.2 | NP_001032370.1 | ||
| NM_053016.5 | 1 | 135 | NP_443749.5 | |
| AJ312216.1 | CAC59693.1 | |||
| AK095003.1 | BAC04472.1 | |||
| BC039306.1 | AAH39306.2 |
Target Gene Details
Entrez Gene ID: | 445815 |
Gene Name: | PALM2-AKAP2 readthrough |
Gene Aliases: |
AKAP2 |
Location: |
Chr.9:109780297-110172512 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PALM2-AKAP2 | NM_007203.4 | 1 | 135 | NP_009134.1 |
| NM_147150.2 | 1 | 135 | NP_671492.1 | |
| AB023137.1 | 1 | 135 | ||
| AJ303079.1 | 1 | 123 | CAC38839.1 | |
| AJ312217.1 | CAC59702.1 | |||
| BC140818.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428222 | Chr.9:109710738 - 109878614 on Build GRCh38 | Loss |
|
| nsv831688 | Chr.9:109776624 - 109936798 on Build GRCh38 | Gain+Loss |
|
| nsv831687 | Chr.9:109738584 - 109893281 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map