Assay Details
Target Gene Details
Entrez Gene ID: | 147841 |
Gene Name: | SPC24, NDC80 kinetochore complex component |
Gene Aliases: |
SPBC24 |
Location: |
Chr.19:11145494-11155997 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPC24 | NM_001317031.1 | 1 | 42 | NP_001303960.1 |
| NM_001317032.1 | 1 | 42 | NP_001303961.1 | |
| NM_001317033.1 | 1 | 42 | NP_001303962.1 | |
| NM_182513.3 | 1 | 42 | NP_872319.1 | |
| XM_005259753.3 | 1 | 227 | XP_005259810.1 | |
| XM_011527702.1 | XP_011526004.1 | |||
| AK075287.1 | BAC11523.1 | |||
| AK303157.1 | ||||
| AK303266.1 | ||||
| AY456387.1 | AAR88651.1 | |||
| BC105037.1 | 1 | 38 | AAI05038.1 | |
| BC105039.1 | 1 | 38 | AAI05040.1 | |
| BQ061822.1 | ||||
| BQ216591.1 | ||||
| HY082333.1 | 1 | 45 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065030 | Chr.19:11149372 - 11199804 on Build GRCh38 | Gain |
|
| nsv833751 | Chr.19:11006139 - 11164225 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map