Assay Details
Target Gene Details
Entrez Gene ID: | 53905 |
Gene Name: | dual oxidase 1 |
Gene Aliases: |
LNOX1, NOXEF1, THOX1 |
Location: |
Chr.15:45129994-45165578 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUOX1 | NM_017434.4 | NP_059130.2 | ||
| NM_175940.2 | NP_787954.1 | |||
| XM_011521681.2 | XP_011519983.1 | |||
| XM_011521682.1 | XP_011519984.1 | |||
| AF213465.1 | AAF71295.1 | |||
| AF230495.1 | AAF73921.1 | |||
| AK075268.1 | ||||
| AK128591.1 | ||||
| AK290372.1 | ||||
| BC114628.1 | ||||
| BC114938.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518213 | Chr.15:45129560 - 45172570 on Build GRCh38 | Gain |
|
| nsv569312 | Chr.15:45089800 - 45168224 on Build GRCh38 | Loss |
|
| nsv569314 | Chr.15:45128520 - 45158125 on Build GRCh38 | Loss |
|
| nsv569313 | Chr.15:45121343 - 45165260 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map