Assay Details
Target Gene Details
Entrez Gene ID: | 51073 |
Gene Name: | mitochondrial ribosomal protein L4 |
Gene Aliases: |
CGI-28, L4mt |
Location: |
Chr.19:10251964-10260060 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL4 | NM_015956.2 | NP_057040.2 | ||
| NM_146387.1 | NP_666499.1 | |||
| NM_146388.1 | 8 | 2019 | NP_666500.1 | |
| AB049635.1 | BAB40840.1 | |||
| AF132962.1 | AAD27737.1 | |||
| AI092267.1 | ||||
| AL561917.2 | 1 | 351 | ||
| AY007152.1 | AAG02005.1 | |||
| BC009858.2 | AAH09858.1 | |||
| BM550378.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74457164,rs77884668] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map