Assay Details
Target Gene Details
Entrez Gene ID: | 80004 |
Gene Name: | epithelial splicing regulatory protein 2 |
Gene Aliases: |
RBM35B |
Location: |
Chr.16:68228547-68236233 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESRP2 | NM_024939.2 | 1 | 345 | NP_079215.2 |
| XM_005256153.4 | 1 | 287 | XP_005256210.1 | |
| XM_006721273.3 | XP_006721336.1 | |||
| AK025571.1 | 1 | 244 | BAB15173.1 | |
| BC030146.2 | 1 | 167 | AAH30146.1 | |
| BU943912.1 | 1 | 179 | ||
| LN680438.1 | 1 | 157 | ||
| LN680439.1 | 1 | 157 |
More Information
Set Membership: |
|

Genomic Map