Assay Details
Target Gene Details
Entrez Gene ID: | 1580 |
Gene Name: | cytochrome P450 family 4 subfamily B member 1 |
Gene Aliases: |
CYPIVB1, P-450HP |
Location: |
Chr.1:46798995-46819349 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP4B1 | NM_000779.3 | 12 | 1735 | NP_000770.2 |
| NM_001099772.1 | 12 | 1738 | NP_001093242.1 | |
| NM_001319161.1 | 11 | 1693 | NP_001306090.1 | |
| NM_001319162.1 | 10 | 1551 | NP_001306091.1 | |
| NM_001319163.1 | 10 | 1548 | NP_001306092.1 | |
| NR_135003.1 | 12 | 1665 | ||
| XM_011540833.1 | 11 | 1371 | XP_011539135.1 | |
| XM_017000466.1 | 11 | 1642 | XP_016855955.1 | |
| AK225576.1 | 12 | 1687 | ||
| BC017758.1 | 12 | 1690 | AAH17758.1 | |
| BM977596.1 | 1 | 436 | ||
| BM982575.1 | 1 | 436 | ||
| CD013981.1 | 12 | 1599 | ||
| J02871.1 | 12 | 1663 | AAA35712.1 | |
| X16699.1 | 12 | 1651 | CAA34672.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585939 | Chr.1:46798765 - 46837514 on Build GRCh38 | Gain |
|
| esv3585938 | Chr.1:46730796 - 46989113 on Build GRCh38 | Gain |
|
| nsv1011086 | Chr.1:46793406 - 46839519 on Build GRCh38 | Gain |
|
| nsv1010063 | Chr.1:46798533 - 46819446 on Build GRCh38 | Gain |
|
| dgv160n100 | Chr.1:46805270 - 46856310 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map