Assay Details
Target Gene Details
Entrez Gene ID: | 134510 |
Gene Name: | ubiquitin like domain containing CTD phosphatase 1 |
Gene Aliases: |
CPUB1 |
Location: |
Chr.5:159263081-159286040 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBLCP1 | NM_145049.3 | NP_659486.2 | ||
| AK057996.1 | BAB71628.1 | |||
| AY444562.1 | AAS68538.1 | |||
| BC013425.2 | AAH13425.2 |
More Information
Set Membership: |
|

Genomic Map