Assay Details
Target Gene Details
Entrez Gene ID: | 79705 |
Gene Name: | leucine rich repeat kinase 1 |
Gene Aliases: |
RIPK6, Roco1 |
Location: |
Chr.15:100919017-101070837 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRK1 | NM_024652.4 | 2 | 362 | NP_078928.3 |
| XM_011522012.2 | 4 | 636 | XP_011520314.1 | |
| XM_011522013.2 | 2 | 345 | XP_011520315.1 | |
| XM_011522014.2 | 2 | 560 | XP_011520316.1 | |
| XM_011522016.2 | XP_011520318.1 | |||
| XM_011522019.2 | 2 | 560 | XP_011520321.1 | |
| AW592808.1 | 2 | 362 | ||
| BC068080.1 | 2 | 232 | AAH68080.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1041172 | Chr.15:100435008 - 101062634 on Build GRCh38 | Gain |
|
| nsv469824 | Chr.15:100759595 - 100934123 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map