Assay Details
Target Gene Details
Entrez Gene ID: | 338328 |
Gene Name: | glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1 |
Gene Aliases: |
GPI-HBP1, HYPL1D |
Location: |
Chr.8:143213193-143217170 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPIHBP1 | NM_001301772.1 | NP_001288701.1 | ||
| NM_178172.5 | 4 | 988 | NP_835466.2 | |
| AY245915.1 | 4 | 988 | AAO86519.1 | |
| BC035810.1 | 4 | 988 | AAH35810.2 | |
| BC063857.1 | 4 | 916 | AAH63857.1 | |
| HY047963.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6438 | Chr.8:143194984 - 143230416 on Build GRCh38 | Insertion |
|
| nsv612751 | Chr.8:143211426 - 143219176 on Build GRCh38 | Gain |
|
| nsv612726 | Chr.8:143147686 - 143256883 on Build GRCh38 | Loss |
|
| nsv465992 | Chr.8:143184697 - 143253125 on Build GRCh38 | Loss |
|
| nsv41 | Chr.8:143202119 - 143230416 on Build GRCh38 | Insertion |
|
| nsv950009 | Chr.8:143207697 - 143235630 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72691630] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map