Assay Details
Target Gene Details
Entrez Gene ID: | 100287944 |
Gene Name: | uncharacterized LOC100287944 |
Gene Aliases: |
- |
Location: |
Chr.12:106496410-106774831 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100287944 | NR_040246.1 | |||
| DA250223.1 |
Target Gene Details
Entrez Gene ID: | 5992 |
Gene Name: | regulatory factor X4 |
Gene Aliases: |
NYD-SP10 |
Location: |
Chr.12:106582907-106762804 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFX4 | NM_213594.2 | NP_998759.1 | ||
| AB095366.1 | BAE48232.1 | |||
| AK291445.1 | ||||
| AY102009.1 | AAM52484.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832511 | Chr.12:106551876 - 106714050 on Build GRCh38 | Gain |
|
| nsv952470 | Chr.12:106579123 - 106587622 on Build GRCh38 | Deletion |
|
| nsv560090 | Chr.12:106541358 - 106830191 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map