Assay Details
Target Gene Details
Entrez Gene ID: | 81532 |
Gene Name: | MOB kinase activator 2 |
Gene Aliases: |
HCCA2 |
Location: |
Chr.11:1469448-1486779 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOB2 | NM_001172223.2 | 5 | 922 | NP_001165694.1 |
| NM_053005.5 | 4 | 709 | NP_443731.2 | |
| AB028173.1 | 4 | 658 | BAB18568.1 | |
| AB066564.1 | 5 | 766 | BAB62266.1 | |
| AJ580639.1 | 4 | 606 | CAE45271.1 | |
| AK057350.1 | 4 | 642 | BAB71443.1 | |
| AK296658.1 | 5 | 889 | ||
| AK300235.1 | 4 | 624 | ||
| BC047291.1 | 4 | 646 | AAH47291.1 | |
| BC067785.1 | 4 | 670 | AAH67785.1 | |
| BX537373.1 | 2 | 2636 | CAD97615.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951279 | Chr.11:1337871 - 1491070 on Build GRCh38 | Deletion |
|
| esv2669189 | Chr.11:864834 - 1489637 on Build GRCh38 | Deletion |
|
| dgv10n68 | Chr.11:1363751 - 1500210 on Build GRCh38 | Loss |
|
| esv2743878 | Chr.11:864732 - 1489652 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map