Assay Details
Target Gene Details
Entrez Gene ID: | 3001 |
Gene Name: | granzyme A |
Gene Aliases: |
CTLA3, HFSP |
Location: |
Chr.5:55102646-55110252 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GZMA | NM_006144.3 | NP_006135.1 | ||
| BC015739.1 | AAH15739.1 | |||
| CR456968.1 | CAG33249.1 | |||
| M18737.1 | AAA52647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4833 | Chr.5:55082169 - 55126059 on Build GRCh38 | Deletion |
|
| nsv830301 | Chr.5:54930046 - 55127677 on Build GRCh38 | Gain |
|
| nsv830302 | Chr.5:55107550 - 55259351 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map