Assay Details
Target Gene Details
Entrez Gene ID: | 51696 |
Gene Name: | hdc homolog, cell cycle regulator |
Gene Aliases: |
HDC, HDCL, HHDC, dJ225E12.1 |
Location: |
Chr.6:139135112-139180809 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HECA | NM_016217.2 | 2 | 1477 | NP_057301.1 |
| AB033492.1 | 2 | 1477 | BAA85322.1 | |
| BC070068.1 | 2 | 1485 | AAH70068.1 |
Target Gene Details
Entrez Gene ID: | 167838 |
Gene Name: | taxilin beta |
Gene Aliases: |
C6orf198, LST001, MDP77, dJ522B19.2 |
Location: |
Chr.6:139159160-139324011 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TXLNB |
More Information
Set Membership: |
|

Genomic Map