Assay Details
Target Gene Details
Entrez Gene ID: | 50863 |
Gene Name: | neurotrimin |
Gene Aliases: |
HNT, IGLON2, NTRI |
Location: |
Chr.11:131370326-132336822 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTM | NM_001048209.1 | NP_001041674.1 | ||
| XM_005271580.3 | XP_005271637.1 | |||
| XM_005271582.3 | XP_005271639.1 | |||
| XM_006718855.3 | XP_006718918.1 | |||
| XM_017017852.1 | XP_016873341.1 | |||
| XM_017017853.1 | XP_016873342.1 | |||
| XM_017017855.1 | XP_016873344.1 | |||
| XM_017017856.1 | XP_016873345.1 | |||
| XM_017017857.1 | XP_016873346.1 | |||
| XM_017017858.1 | XP_016873347.1 | |||
| XM_017017859.1 | XP_016873348.1 | |||
| XM_017017861.1 | XP_016873350.1 | |||
| XM_017017862.1 | XP_016873351.1 | |||
| XM_017017865.1 | XP_016873354.1 | |||
| AK293487.1 | ||||
| AK294137.1 | ||||
| AK315964.1 | ||||
| AY358331.1 | AAQ88697.1 |
Target Gene Details
Entrez Gene ID: | 101929637 |
Gene Name: | NTM antisense RNA 1 |
Gene Aliases: |
C11orf39 |
Location: |
Chr.11:131662129-131663556 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTM-AS1 | NR_126159.1 | |||
| AK127362.1 | 1 | 181 | BAC86944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1036099 | Chr.11:131643993 - 131718467 on Build GRCh38 | Loss |
|
| nsv428266 | Chr.11:131562907 - 131879091 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map