Assay Details
Target Gene Details
Entrez Gene ID: | 4848 |
Gene Name: | CCR4-NOT transcription complex subunit 2 |
Gene Aliases: |
CDC36, HSPC131, NOT2, NOT2H |
Location: |
Chr.12:70242993-70354993 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT2 | NM_001199302.1 | 2 | 318 | NP_001186231.1 |
| NM_014515.5 | 1 | 449 | NP_055330.1 | |
| XM_006719429.3 | 1 | 450 | XP_006719492.1 | |
| XM_006719430.3 | 1 | 450 | XP_006719493.1 | |
| XM_006719432.3 | 1 | 450 | XP_006719495.1 | |
| XM_006719433.3 | 1 | 450 | XP_006719496.1 | |
| XM_011538400.2 | 1 | 450 | XP_011536702.1 | |
| XM_017019348.1 | 1 | 450 | XP_016874837.1 | |
| XM_017019349.1 | 1 | 450 | XP_016874838.1 | |
| XM_017019350.1 | 1 | 450 | XP_016874839.1 | |
| XM_017019351.1 | 1 | 450 | XP_016874840.1 | |
| XM_017019352.1 | 1 | 450 | XP_016874841.1 | |
| XM_017019353.1 | 1 | 450 | XP_016874842.1 | |
| XM_017019354.1 | 1 | 450 | XP_016874843.1 | |
| AF044215.1 | AAQ13426.1 | |||
| AF161480.1 | AAF29095.1 | |||
| AF180473.1 | AAF29827.1 | |||
| AK000662.1 | BAA91313.1 | |||
| AK315715.1 | 2 | 319 | ||
| BC002597.2 | AAH02597.1 | |||
| BC011826.2 | AAH11826.1 | |||
| DA445519.1 | 1 | 446 | ||
| DB038192.1 | 1 | 449 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3629884 | Chr.12:70233284 - 70252849 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map