Assay Details
Target Gene Details
Entrez Gene ID: | 2904 |
Gene Name: | glutamate ionotropic receptor NMDA type subunit 2B |
Gene Aliases: |
EIEE27, GluN2B, MRD6, NMDAR2B, NR2B, hNR3 |
Location: |
Chr.12:13537337-13982012 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIN2B | NM_000834.3 | 13 | 3488 | NP_000825.2 |
| XM_005253351.3 | 4 | 1215 | XP_005253408.1 | |
| XM_011520628.2 | 14 | 4281 | XP_011518930.1 | |
| XM_011520629.2 | 15 | 4088 | XP_011518931.1 | |
| XM_017019219.1 | 14 | 3836 | XP_016874708.1 | |
| AB208850.1 | 10 | 2267 | BAD92087.1 | |
| BC113618.1 | 13 | 3387 | ||
| BC113620.1 | 13 | 3387 | ||
| U11287.1 | 13 | 3519 | AAB60368.1 | |
| U88963.1 | 13 | 3488 | AAD00659.1 | |
| U90278.1 | 13 | 3518 | AAB49993.1 |
More Information
Set Membership: |
|

Genomic Map