Assay Details
Target Gene Details
Entrez Gene ID: | 5689 |
Gene Name: | proteasome subunit beta 1 |
Gene Aliases: |
HC5, PMSB1, PSC5 |
Location: |
Chr.6:170535116-170553329 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMB1 | NM_002793.3 | 4 | 466 | NP_002784.1 |
| AB209078.1 | 4 | 396 | BAD92315.1 | |
| AB451312.1 | 4 | 379 | ||
| AB451442.1 | 4 | 379 | ||
| AK023290.1 | 3 | 1594 | ||
| AK058050.1 | 4 | 436 | ||
| AK223193.1 | 4 | 417 | BAD96913.1 | |
| BC000508.2 | 4 | 417 | AAH00508.1 | |
| BC020807.1 | 4 | 417 | AAH20807.1 | |
| BP369175.1 | 4 | 478 | ||
| BQ685374.1 | 3 | 196 | ||
| BT019720.1 | 4 | 379 | AAV38525.1 | |
| D00761.1 | 4 | 391 | BAA00658.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428158 | Chr.6:170272663 - 170700831 on Build GRCh38 | Gain |
|
| nsv1015584 | Chr.6:170515957 - 170579200 on Build GRCh38 | Gain |
|
| nsv1033491 | Chr.6:170342284 - 170673434 on Build GRCh38 | Loss |
|
| nsv830874 | Chr.6:170538725 - 170729166 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map