Assay Details
Target Gene Details
Entrez Gene ID: | 2267 |
Gene Name: | fibrinogen like 1 |
Gene Aliases: |
HFREP1, HP-041, LFIRE-1, LFIRE1 |
Location: |
Chr.8:17864380-17909983 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGL1 | NM_004467.3 | 8 | 966 | NP_004458.3 |
| NM_147203.2 | 8 | 1054 | NP_671736.2 | |
| NM_201552.1 | 9 | 1093 | NP_963846.1 | |
| NM_201553.1 | 9 | 1181 | NP_963847.1 | |
| XM_017013232.1 | 9 | 1943 | XP_016868721.1 | |
| XM_017013233.1 | 7 | 1018 | XP_016868722.1 | |
| AF042354.1 | 8 | 1002 | ||
| AF168954.1 | 8 | 989 | AAM88911.1 | |
| AK225616.1 | 8 | 952 | ||
| AK225619.1 | 8 | 941 | ||
| AK289448.1 | 8 | 952 | ||
| AL514837.3 | 1 | 290 | ||
| AX772838.1 | 9 | 1046 | CAE06268.1 | |
| BC007047.1 | 8 | 962 | AAH07047.1 | |
| BT006635.1 | 7 | 857 | AAP35281.1 | |
| D14446.1 | 8 | 935 | BAA03336.1 | |
| D87342.1 | 7 | 862 | BAB70690.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831250 | Chr.8:17737238 - 17890332 on Build GRCh38 | Loss |
|
| dgv7121n100 | Chr.8:17766079 - 17957752 on Build GRCh38 | Gain |
|
| nsv610701 | Chr.8:17787519 - 17956578 on Build GRCh38 | Gain |
|
| esv3616500 | Chr.8:17771011 - 17959017 on Build GRCh38 | Gain |
|
| esv2762735 | Chr.8:17723646 - 17957764 on Build GRCh38 | Gain |
|
| nsv1024843 | Chr.8:17813558 - 17869789 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map