Assay Details
Target Gene Details
Entrez Gene ID: | 10322 |
Gene Name: | SMYD family member 5 |
Gene Aliases: |
NN8-4AG, RAI15, RRG1, ZMYND23 |
Location: |
Chr.2:73214238-73227227 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMYD5 | NM_006062.2 | 13 | 1295 | NP_006053.2 |
| XM_006711918.3 | 13 | 1382 | XP_006711981.1 | |
| XM_017003163.1 | 12 | 3015 | XP_016858652.1 | |
| AK300779.1 | 13 | 1269 | ||
| AK301809.1 | 13 | 1288 | ||
| BC073806.1 | 13 | 1271 | AAH73806.1 | |
| U50383.1 | 13 | 1248 | AAB38131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2776 | Chr.2:73204726 - 73255176 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map