Assay Details
Target Gene Details
Entrez Gene ID: | 6543 |
Gene Name: | solute carrier family 8 member A2 |
Gene Aliases: |
NCX2 |
Location: |
Chr.19:47428017-47472050 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC8A2 | NM_015063.2 | NP_055878.1 | ||
| XM_005259172.1 | XP_005259229.1 | |||
| XM_017027159.1 | XP_016882648.1 | |||
| AB029010.1 | ||||
| AK293457.1 | ||||
| AK293836.1 | ||||
| AK293860.1 | ||||
| AK302552.1 | ||||
| AL512764.1 | CAC21681.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428366 | Chr.19:47354499 - 47522248 on Build GRCh38 | Loss |
|
| nsv1057003 | Chr.19:47416492 - 47471403 on Build GRCh38 | Loss |
|
| nsv517901 | Chr.19:47373953 - 47444971 on Build GRCh38 | Loss |
|
| nsv579888 | Chr.19:47428355 - 47658874 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map