Assay Details
Target Gene Details
Entrez Gene ID: | 283748 |
Gene Name: | phospholipase A2 group IVD |
Gene Aliases: |
cPLA2delta |
Location: |
Chr.15:42067562-42094559 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G4D | NM_178034.3 | NP_828848.3 | ||
| XM_011521467.2 | XP_011519769.1 | |||
| AB090876.1 | BAC67158.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035998 | Chr.15:41806698 - 42142056 on Build GRCh38 | Gain |
|
| nsv471683 | Chr.15:42042177 - 42210784 on Build GRCh38 | Gain+Loss |
|
| nsv832985 | Chr.15:42079284 - 42272768 on Build GRCh38 | Loss |
|
| nsv1043641 | Chr.15:42044386 - 42089042 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75621794] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map