Assay Details
Target Gene Details
Entrez Gene ID: | 55897 |
Gene Name: | mesoderm posterior bHLH transcription factor 1 |
Gene Aliases: |
bHLHc5 |
Location: |
Chr.15:89734772-89751309 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MESP1 |
Target Gene Details
Entrez Gene ID: | 56964 |
Gene Name: | WD repeat domain 93 |
Gene Aliases: |
- |
Location: |
Chr.15:89690074-89744390 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR93 | NM_001284395.1 | 15 | 1684 | NP_001271324.1 |
| NM_020212.1 | 15 | 1768 | NP_064597.1 | |
| XM_006720614.1 | 14 | 1690 | XP_006720677.1 | |
| XM_006720615.1 | 13 | 1603 | XP_006720678.1 | |
| XM_011521794.2 | 14 | 1983 | XP_011520096.1 | |
| XM_011521795.1 | 14 | 1714 | XP_011520097.1 | |
| XM_011521796.2 | 14 | 1690 | XP_011520098.1 | |
| XM_011521798.2 | 13 | 1974 | XP_011520100.1 | |
| XM_011521800.1 | 12 | 1144 | XP_011520102.1 | |
| XM_017022415.1 | 13 | 1607 | XP_016877904.1 | |
| XM_017022416.1 | 11 | 1074 | XP_016877905.1 | |
| AK097525.1 | 15 | 1652 | BAC05088.1 | |
| AK302032.1 | 9 | 1173 | ||
| AL390084.1 | 7 | 752 | CAB98208.1 | |
| AL390085.1 | 7 | 752 | CAB98209.1 | |
| BC064626.1 | 15 | 1768 | AAH64626.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv570388 | Chr.15:89730911 - 89794962 on Build GRCh38 | Loss |
|
| nsv1046117 | Chr.15:89683716 - 89765410 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map