Assay Details
Target Gene Details
Entrez Gene ID: | 5319 |
Gene Name: | phospholipase A2 group IB |
Gene Aliases: |
PLA2, PLA2A, PPLA2 |
Location: |
Chr.12:120322111-120327789 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G1B | NM_000928.2 | NP_000919.1 | ||
| AA844927.1 | ||||
| AK311830.1 | ||||
| BC005386.1 | AAH05386.1 | |||
| BC106725.1 | AAI06726.1 | |||
| BC106726.1 | AAI06727.1 | |||
| BX113838.1 | ||||
| CA867923.1 | ||||
| M21054.1 | AAA36450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560435 | Chr.12:120257338 - 120331854 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map