Assay Details
Target Gene Details
Entrez Gene ID: | 5354 |
Gene Name: | proteolipid protein 1 |
Gene Aliases: |
GPM6C, HLD1, MMPL, PLP, PLP/DM20, PMD, SPG2 |
Location: |
Chr.X:103776506-103792619 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLP1 | NM_000533.4 | 7 | 2273 | NP_000524.3 |
| NM_001128834.2 | 8 | 2355 | NP_001122306.1 | |
| NM_001305004.1 | 7 | 2048 | NP_001291933.1 | |
| NM_199478.2 | 7 | 2168 | NP_955772.1 | |
| AK128782.1 | 7 | 2196 | ||
| BC002665.1 | 7 | 2111 | AAH02665.1 | |
| BC095452.1 | 7 | 2157 | AAH95452.1 | |
| EB387051.1 | 1 | 513 | ||
| M27110.1 | 7 | 2188 | AAA60117.1 | |
| M54927.1 | 7 | 2191 | AAA59565.1 |
More Information
Set Membership: |
|

Genomic Map