Assay Details
Target Gene Details
Entrez Gene ID: | 23019 |
Gene Name: | CCR4-NOT transcription complex subunit 1 |
Gene Aliases: |
AD-005, CDC39, NOT1, NOT1H |
Location: |
Chr.16:58519946-58629886 on Build GRCh38 |
Assay Gene Location: | Within Exon 40 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT1 | NM_001265612.1 | 40 | 6053 | NP_001252541.1 |
| NM_016284.4 | 40 | 6068 | NP_057368.3 | |
| NR_049763.1 | 40 | 6053 | ||
| AB023224.2 | 27 | 4113 | BAA76851.2 | |
| AK056927.1 | 17 | 2398 | ||
| AK075125.1 | 10 | 1367 | ||
| AK300281.1 | ||||
| AK302495.1 | 17 | 2438 | ||
| AL117492.1 | 12 | 1907 | CAB55960.1 | |
| AL832769.1 | 40 | 6017 | ||
| AL833549.1 | 40 | 6027 | ||
| BC000779.2 | 2 | 226 | AAH00779.2 | |
| BC024317.1 | 23 | 3467 | AAH24317.1 | |
| BX537840.1 | 40 | 5999 | CAD97851.1 | |
| CR749237.1 | 33 | 5008 | CAH18093.1 | |
| EF553522.1 | 40 | 6002 |
More Information
Set Membership: |
|

Genomic Map