Assay Details
Target Gene Details
Entrez Gene ID: | 3293 |
Gene Name: | hydroxysteroid 17-beta dehydrogenase 3 |
Gene Aliases: |
EDH17B3, SDR12C2 |
Location: |
Chr.9:96235306-96313987 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSD17B3 | NM_000197.1 | NP_000188.1 | ||
| XM_011518618.2 | XP_011516920.1 | |||
| XM_011518619.2 | XP_011516921.1 | |||
| XM_017014671.1 | XP_016870160.1 | |||
| XM_017014672.1 | XP_016870161.1 | |||
| XM_017014673.1 | XP_016870162.1 | |||
| XM_017014674.1 | XP_016870163.1 | |||
| XM_017014675.1 | XP_016870164.1 | |||
| XM_017014677.1 | XP_016870166.1 | |||
| BC034281.1 | AAH34281.1 | |||
| U05659.1 | AAC50066.1 |
More Information
Set Membership: |
|

Genomic Map