Assay Details
Target Gene Details
Entrez Gene ID: | 10524 |
Gene Name: | lysine acetyltransferase 5 |
Gene Aliases: |
ESA1, HTATIP, HTATIP1, PLIP, TIP, TIP60, ZC2HC5, cPLA2 |
Location: |
Chr.11:65711996-65719606 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KAT5 | NM_001206833.1 | 12 | 1678 | NP_001193762.1 |
| NM_006388.3 | 14 | 1735 | NP_006379.2 | |
| NM_182709.2 | 13 | 1579 | NP_874368.1 | |
| NM_182710.2 | 13 | 1834 | NP_874369.1 | |
| XM_006718421.2 | 14 | 1768 | XP_006718484.1 | |
| AB209813.1 | 11 | 1381 | BAD93050.1 | |
| AK225100.1 | ||||
| AK226002.1 | ||||
| AK304664.1 | 12 | 1428 | ||
| AK313297.1 | 14 | 1711 | ||
| BC000166.3 | 12 | 1301 | AAH00166.3 | |
| BC064912.1 | 14 | 1475 | AAH64912.1 | |
| BC093032.1 | 13 | 1346 | AAH93032.1 | |
| BC117167.1 | 14 | 1522 | ||
| BC143296.1 | 13 | 1363 | ||
| BU737532.1 | ||||
| U40989.1 | 13 | 1440 | AAB02683.1 | |
| U67734.1 | 13 | 1330 | AAD00163.1 | |
| U74667.1 | 14 | 1710 | AAB18236.1 |
Target Gene Details
Entrez Gene ID: | 84153 |
Gene Name: | ribonuclease H2 subunit C |
Gene Aliases: |
AGS3, AYP1 |
Location: |
Chr.11:65717673-65720938 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASEH2C | NM_032193.3 | 4 | 1350 | NP_115569.2 |
More Information
Set Membership: |
|

Genomic Map